Learn More
SSEA-1 Antibody (MC-480), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand:  Novus Biologicals NB100-1831SS
 View more versions of this product
Product Code. 18009223
  • £186.00 / 25µL
Description
SSEA-1 Monoclonal specifically detects SSEA-1 in Human, Mouse, Rat, Chimpanzee samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SSEA-1
Monoclonal
1 mg/mL
Western Blot 2 ug/ml, Simple Western 1:10, Flow Cytometry 2.5 ug/million cells, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:50-1:200
alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1
Mouse
IgM purified
RUO
Primary
Human and mouse. This antibody recognizes SSEA1 in mouse EC and ES cells but not human, but SSEA1 is expressed by some human differentiated cell types (ie: trophoblasts). Reactivity with Rat is reported by Kim et al. 2007 (PMID 17548530).
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgM
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
MC-480
Unconjugated
PBS with 0.02% Sodium Azide
FUT4
F9 tetracarcinoma stem cells (X-irradiated).
0.025 mL
Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells
2526
Human, Mouse, Rat, Chimpanzee
Purified