missing translation for 'onlineSavingsMsg'
Learn More

SS18 Antibody (SS18.SSX/12628R), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-44631-100ug

 View more versions of this product

Product Code. 30289844

  • £392.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SS18 Monoclonal antibody specifically detects SS18 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SS18
Monoclonal
0.2 mg/ml
10 mM PBS with 0.05% BSA
Rabbit
Protein A or G purified
RUO
Primary
Human
Purified
Immunohistochemistry, Western Blot
SS18.SSX/12628R
Unconjugated
MGC116875, Protein SYT, SSXTsynovial sarcoma, translocated to X chromosome, Synovial sarcoma translocated to X chromosome protein, synovial sarcoma translocation, chromosome 18, SYTprotein SSXT
Recombinant fragment corresponding to the fusion site of human SS18 protein (Uniprot: Q15532). Exact sequence is proprietary.
100 μg
Core ESC Like Genes, Stem Cell Markers
6760
Store at 4⪚C.
IgG κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.