missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SS18 Antibody (SS18.SSX/12628R), Novus Biologicals™
Rabbit Monoclonal Antibody
£219.00 - £392.00
Specifications
| Antigen | SS18 |
|---|---|
| Clone | SS18.SSX/12628R |
| Concentration | 0.2 mg/ml |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30290090
|
Novus Biologicals
NBP3-44631-20ug |
20 μg |
£219.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30289844
|
Novus Biologicals
NBP3-44631-100ug |
100 μg |
£392.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SS18 Monoclonal antibody specifically detects SS18 in Human samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| SS18 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC116875, Protein SYT, SSXTsynovial sarcoma, translocated to X chromosome, Synovial sarcoma translocated to X chromosome protein, synovial sarcoma translocation, chromosome 18, SYTprotein SSXT | |
| Recombinant fragment corresponding to the fusion site of human SS18 protein (Uniprot: Q15532). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
| SS18.SSX/12628R | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 6760 | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title