missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRRM2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | SRRM2 |
|---|---|
| Concentration | 300 μg/mL |
| Applications | Immunofluorescence, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18010536
|
Proteintech
30741-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18000516
|
Proteintech
30741-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SRRM2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SRRM2. This antibody recognizes human antigen. The SRRM2 antibody has been validated for the following applications: IF, ELISA analysis.
Specifications
| SRRM2 | |
| Immunofluorescence, ELISA | |
| Rabbit | |
| 23524 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 300 μg/mL | |
| Liquid | |
| Human | |
| 300 KD, 300 kDa nuclear matrix antigen, CWF21, DKFZp686O15166, FLJ21926, FLJ22250, KIAA0324, SRL300, SRm300, SRRM2, TaxREB803 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title