Learn More
SRRM2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30741-1-AP-150UL
This item is not returnable.
View return policy
Description
The SRRM2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SRRM2. This antibody recognizes human antigen. The SRRM2 antibody has been validated for the following applications: IF, ELISA analysis.
Specifications
| SRRM2 | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Immunofluorescence, ELISA | |
| 300 KD, 300 kDa nuclear matrix antigen, CWF21, DKFZp686O15166, FLJ21926, FLJ22250, KIAA0324, SRL300, SRm300, SRRM2, TaxREB803 | |
| 150 μL | |
| 23524 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |