missing translation for 'onlineSavingsMsg'
Learn More

SRP9 Mouse anti-Human, Mouse, Rat, Clone: 1H7G10, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66068-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16867605

  • £339.00 / 150µL
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. SRP9 together with SRP14 and the Alu portion of the SRP RNA, constitutes the elongation arrest domain of SRP. The complex of SRP9 and SRP14 is required for SRP RNA binding.
Specifications

Specifications

SRP9
Monoclonal
0.93 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P49458, P49962
SRP9
SRP9 Fusion Protein Ag17153
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG2a
Western Blot
1H7G10
Unconjugated
SRP9
ALURBP, SRP9
Mouse
Protein A
RUO
27058, 6726, 690345
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers