missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRP9 Mouse anti-Human, Mouse, Rat, Clone: 1H7G10, Proteintech
Mouse Monoclonal Antibody
£130.00 - £339.00
Specifications
Antigen | SRP9 |
---|---|
Clone | 1H7G10 |
Concentration | 0.93 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877605
|
Proteintech
66068-1-IG-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867605
|
Proteintech
66068-1-IG-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. SRP9 together with SRP14 and the Alu portion of the SRP RNA, constitutes the elongation arrest domain of SRP. The complex of SRP9 and SRP14 is required for SRP RNA binding.Specifications
SRP9 | |
0.93 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ALURBP, SRP9 | |
SRP9 | |
IgG2a | |
Protein A | |
Antibody |
1H7G10 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P49458, P49962 | |
27058, 6726, 690345 | |
SRP9 Fusion Protein Ag17153 | |
Primary | |
-20°C | |
SRP9 |