missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRGN Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £366.00
Specifications
| Antigen | SRGN |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18683282
|
Novus Biologicals
NBP2-95229-0.02ml |
0.02 mL |
£149.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18636132
|
Novus Biologicals
NBP2-95229-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SRGN Polyclonal antibody specifically detects SRGN in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SRGN | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Biology, Cytoskeleton Markers, Diabetes Research, Endocrinology, metabolism, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 5552 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ12930, Hematopoetic proteoglycan core protein, MGC9289, P.PG, Platelet proteoglycan core protein, platelet proteoglycan protein core, PPG, PRG1p.PG, PRGhematopoetic proteoglycan core peptide, proteoglycan 1, secretory granule, proteoglycan protein core for mast cell secretory granule, secretory granule proteoglycan 1, secretory granule proteoglycan core peptide, Secretory granule proteoglycan core protein, serglycin, serglycin proteoglycan | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 28-158 of human SRGN (NP_002718.2). YPTRRARYQWVRCNPDSNSANCLEEKGPMFELLPGESNKIPRLRTDLFPKTRIQDLNRIFPLSEDYSGSGFGSGSGSGSGSGSGFLTEMEQDYQLVDESDAFHDNLRSLDRNLPSDSQDLGQHGLEEDFML | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title