missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRGN Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95229-0.02ml
This item is not returnable.
View return policy
Description
SRGN Polyclonal antibody specifically detects SRGN in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SRGN | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| FLJ12930, Hematopoetic proteoglycan core protein, MGC9289, P.PG, Platelet proteoglycan core protein, platelet proteoglycan protein core, PPG, PRG1p.PG, PRGhematopoetic proteoglycan core peptide, proteoglycan 1, secretory granule, proteoglycan protein core for mast cell secretory granule, secretory granule proteoglycan 1, secretory granule proteoglycan core peptide, Secretory granule proteoglycan core protein, serglycin, serglycin proteoglycan | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 28-158 of human SRGN (NP_002718.2). YPTRRARYQWVRCNPDSNSANCLEEKGPMFELLPGESNKIPRLRTDLFPKTRIQDLNRIFPLSEDYSGSGFGSGSGSGSGSGSGFLTEMEQDYQLVDESDAFHDNLRSLDRNLPSDSQDLGQHGLEEDFML | |
| 0.02 mL | |
| Apoptosis, Cell Biology, Cytoskeleton Markers, Diabetes Research, Endocrinology, metabolism, Signal Transduction | |
| 5552 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction