missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testican 3/SPOCK3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59703
This item is not returnable.
View return policy
Description
Testican 3/SPOCK3 Polyclonal specifically detects Testican 3/SPOCK3 in Human samples. It is validated for Western Blot.
Specifications
| Testican 3/SPOCK3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CWCV, and Kazal-like domains proteoglycan 3, HSAJ1454, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3, TES-3, testican 3, TICN3 | |
| Rabbit | |
| 49 kDa | |
| 100 μL | |
| Signal Transduction | |
| 50859 | |
| Human, Mouse, Rat, Pig, Canine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9BQ16-1 | |
| SPOCK3 | |
| Synthetic peptides corresponding to SPOCK3(sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3) The peptide sequence was selected from the middle region of SPOCK3. Peptide sequence VDRYGNEVMGSRINGVADCAIDFEISGDFASGDFHEWTDDEDDEDDIMND The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction