missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testican 3/SPOCK3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | Testican 3/SPOCK3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Testican 3/SPOCK3 Polyclonal specifically detects Testican 3/SPOCK3 in Human samples. It is validated for Western Blot.Specifications
| Testican 3/SPOCK3 | |
| Polyclonal | |
| Rabbit | |
| Q9BQ16-1 | |
| 50859 | |
| IgG | |
| 49 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| CWCV, and Kazal-like domains proteoglycan 3, HSAJ1454, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3, TES-3, testican 3, TICN3 | |
| SPOCK3 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title