missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRBD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57557
This item is not returnable.
View return policy
Description
SRBD1 Polyclonal specifically detects SRBD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SRBD1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FLJ10379, H_NH0576F01.1, S1 RNA binding domain 1, S1 RNA-binding domain-containing protein 1, WUGSC:H_NH0576F01.1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55133 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 4.0-8.0 ug/ml, Immunohistochemistry-Paraffin | |
| Q8N5C6 | |
| SRBD1 | |
| Synthetic peptides corresponding to SRBD1(S1 RNA binding domain 1) The peptide sequence was selected from the N terminal of SRBD1. Peptide sequence MSSLPRRAKVQVQDVVLKDEFSSFSELSSASEEDDKEDSAWEPQKKVPRS. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Equine: 100%; Human: 100%; Rabbit: 100%; Bovine: 92%; Mouse: 85%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit, Yeast, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction