missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-BI/SR-BII Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 9 publications
£183.00 - £391.00
Specifications
| Antigen | SR-BI/SR-BII |
|---|---|
| Dilution | Western Blot 1:1000, Flow Cytometry 1:400. Use reported in scientific literature (PMID 22097902), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative) 5 ug/ml, Knockout Validated, Block/Neutralize reported in scientific literature (PMID 24859737) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Neutralization |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18116939
|
Novus Biologicals
NB400-134SS |
0.025 mL |
£183.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18798453
|
Novus Biologicals
NB400-134 |
£391.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
SR-BI/SR-BII Polyclonal specifically detects SR-BI/SR-BII in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative), Knockout Validated, Block/Neutralize.Specifications
| SR-BI/SR-BII | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Neutralization | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| Whole antisera with 0.02% Sodium Azide | |
| CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1scavenger receptor class B type 1, CLA1CD36 antigen-like 1, CLA-1HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, scavenger receptor class B type III, scavenger receptor class B, member 1, SRB1scavenger receptor class B member 1, SR-BIMGC138242 | |
| SCARB1 | |
| IgG | |
| Unpurified | |
| Mouse. Previous lots have been shown to react with rat and human as well. |
| Western Blot 1:1000, Flow Cytometry 1:400. Use reported in scientific literature (PMID 22097902), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative) 5 ug/ml, Knockout Validated, Block/Neutralize reported in scientific literature (PMID 24859737) | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| Q61009, O35114 | |
| 949 | |
| A peptide from the extracellular domain (residues 230-380) of Scavenger Receptor-BI/BII that was expressed as two tandem copies in bacteria using the pET system. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 82 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title