Learn More
SR-BI/SR-BII Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 9 publications
Brand: Novus Biologicals NB400-134
This item is not returnable.
View return policy
Description
SR-BI/SR-BII Polyclonal specifically detects SR-BI/SR-BII in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative), Knockout Validated, Block/Neutralize.
Specifications
| 82 kDa | |
| Unpurified | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| Antisera |