Learn More
Gilac™ Square Bucket with Watertight Lid, 12 L
Product Code. 17998511
Change view
Click to view available options
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
17998511 Each
17908861 Each
17998431 Each
17978671 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17998511 Supplier Gilac™ Supplier No. G615712
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Square 12 L Bucket with Watertight Lid fit for food contact with watertight lid and pouring spout.
  • Bucket fit for food contact with watertight lid and pouring spout.
  • Airtight lid keeps flavours in and odours out.
  • Nesting when empty, no suction and stacking when lid is closed.
  • Enables liquid preparations to be stored and transported.
  • Graduation in litres.
  • Discharge area that prevents water retention when being cleaned and anti-wear skirt that lifts the bucket off the ground.
  • HDPE body and LDPE lid.
  • Stainless steel handle.
  • External dimensions include handle.
Specifications
Includes Bucket, Handle, and Lid
Quantity 12 L