missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SQLE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12544-1-AP-20UL
This item is not returnable.
View return policy
Description
SQLE, also named as ERG1, SE and SM, belongs to the squalene monooxygenase family. It catalyzes the first oxygenation step in cholesterol synthesis, acting on squalene before cyclization into the basic steroid structure. SQLE may serve as a flux-controlling enzyme beyond 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMGR, considered as rate limiting). It is also posttranslationally regulated by cholesterol-dependent proteasomal degradation. The MW of SQLE is about 64kd.Specifications
| SQLE | |
| Polyclonal | |
| Unconjugated | |
| SQLE | |
| ERG1, SE, SM, SQLE, squalene epoxidase, Squalene monooxygenase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20775, 29230, 6713 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P52019, P52020, Q14534 | |
| SQLE | |
| SQLE Fusion Protein Ag3266 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction