Learn More
SQLE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12544-1-AP-150UL
 View more versions of this product
Product Code. 16820304
  • £349.00 / 150µL
Description
SQLE, also named as ERG1, SE and SM, belongs to the squalene monooxygenase family. It catalyzes the first oxygenation step in cholesterol synthesis, acting on squalene before cyclization into the basic steroid structure. SQLE may serve as a flux-controlling enzyme beyond 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMGR, considered as rate limiting). It is also posttranslationally regulated by cholesterol-dependent proteasomal degradation. The MW of SQLE is about 64kd.
Specifications
Specifications
SQLE
Polyclonal
Unconjugated
SQLE
ERG1, SE, SM, SQLE, squalene epoxidase, Squalene monooxygenase
Rabbit
Antigen Affinity Chromatography
RUO
20775, 29230, 6713
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P52019, P52020, Q14534
SQLE
SQLE Fusion Protein Ag3266
150 μL
Primary
Human, Mouse, Rat