missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPTLC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
£178.00 - £432.00
Specifications
| Antigen | SPTLC1 |
|---|---|
| Concentration | 0.94 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17740814
![]() |
Invitrogen™
PA5119409 |
200 μL |
£432.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17753244
|
Invitrogen™
PA5121847 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPTLC1 Polyclonal Antibody for Western Blot, IHC (P)
Serine palmitoyltransferase (SPT) is the key enzyme in sphingolipid biosynthesis. It catalyzes the pyridoxal-5-prime-phosphate-dependent condensation of L-serine and palmitoyl-CoA to 3-oxosphinganine.Specifications
| SPTLC1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| Sptlc1 | |
| AW552086; C77762; E030036H05; fc75h04; fp41c08; hLCB1; HSAN; HSAN1; HSN1; hypothetical protein LOC553981; hypothetical protein LOC614165; LBC1; LCB 1; LCB1; long chain base biosynthesis protein 1; MGC14645; rCG_44191; RGD1306617; RP11-118F2.1; serine C-palmitoyltransferase; serine palmitoyltransferase 1; serine palmitoyltransferase long chain base subunit 1; serine palmitoyltransferase subunit 1; serine palmitoyltransferase, long chain base subunit 1; serine-palmitoyl-CoA transferase 1; SPT 1; Spt1; SPTI; sptlc1; wu:fc75h04; wu:fp41c08; zgc:112247 | |
| Sptlc1 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.94 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O15269, O35704 | |
| 10558, 268656 | |
| Recombinant protein Serine Palmitoyltransferase, Long Chain Base Subunit 1. The antigen corresponds to amino acid range 143-473 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title