missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPTLC1 Mouse anti-Human, Mouse, Rat, CoraLite 594, Clone: 2D11D7, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | SPTLC1 |
|---|---|
| Clone | 2D11D7 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17731439
|
Proteintech
CL594-66899-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17721439
|
Proteintech
CL594-66899-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPTLC1 Monoclonal antibody specifically detects SPTLC1 in Human, Mouse, Rat samples. It is validated for IFSpecifications
| SPTLC1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| 10558 | |
| Primary | |
| Store at -20°C |
| 2D11D7 | |
| Immunofluorescence | |
| CoraLite 594 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| IgG1 | |
| Protein G purification | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title