missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPTLC1 Mouse anti-Human, Mouse, Rat, CoraLite 594, Clone: 2D11D7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66899-100UL
This item is not returnable.
View return policy
Description
SPTLC1 Monoclonal antibody specifically detects SPTLC1 in Human, Mouse, Rat samples. It is validated for IFSpecifications
| SPTLC1 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| Mouse | |
| 100 μL | |
| 10558 | |
| Store at -20°C | |
| Liquid |
| Immunofluorescence | |
| 2D11D7 | |
| CoraLite 594 | |
| BC069298 | |
| Protein G purification | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction