missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPTBN4 Antibody (S393-2), Novus Biologicals™
Mouse Monoclonal Antibody
£168.00 - £427.00
Specifications
| Antigen | SPTBN4 |
|---|---|
| Clone | S393-2 |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18296455
|
Novus Biologicals
NBP2-59321 |
100 μg |
£427.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18695766
|
Novus Biologicals
NBP2-59321-25ug |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPTBN4 Monoclonal specifically detects SPTBN4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| SPTBN4 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 57731 | |
| Fusion protein amino acids 1621-1832 (C-terminal repeats 14 to 15) of human Beta4-spectrin | |
| Primary | |
| Detects >200kDa. Does not cross react with other Beta-spectrins |
| S393-2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| beta-IV spectrin, KIAA1642, QV, spectrin beta chain, brain 3, spectrin, beta, non-erythrocytic 4, spectrin, non-erythroid beta chain 3, SPNB4, SPTBN3 | |
| SPTBN4 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title