missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPTBN4 Antibody (S393-2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59321-25ug
This item is not returnable.
View return policy
Description
SPTBN4 Monoclonal specifically detects SPTBN4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| SPTBN4 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.1% Sodium Azide | |
| SPTBN4 | |
| Fusion protein amino acids 1621-1832 (C-terminal repeats 14 to 15) of human Beta4-spectrin | |
| 25 μg | |
| Primary | |
| Detects >200kDa. Does not cross react with other Beta-spectrins | |
| Store at -20°C. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| S393-2 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| beta-IV spectrin, KIAA1642, QV, spectrin beta chain, brain 3, spectrin, beta, non-erythrocytic 4, spectrin, non-erythroid beta chain 3, SPNB4, SPTBN3 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 57731 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction