missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRY4 Antibody (SR1553), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | SPRY4 |
|---|---|
| Clone | SR1553 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18696104
|
Novus Biologicals
NBP3-21971-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660874
|
Novus Biologicals
NBP3-21971-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPRY4 Monoclonal antibody specifically detects SPRY4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| SPRY4 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| protein sprouty homolog 4, sprouty homolog 4 (Drosophila), spry-4 | |
| A synthesized peptide derived from human SPRY4 (Uniprot #: Q9C004) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| SR1553 | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Growth and Development, Neuronal Cell Markers, Signal Transduction | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| 81848 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title