missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRY4 Antibody (SR1553), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21971-50ul
This item is not returnable.
View return policy
Description
SPRY4 Monoclonal antibody specifically detects SPRY4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| SPRY4 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| SR1553 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| protein sprouty homolog 4, sprouty homolog 4 (Drosophila), spry-4 | |
| A synthesized peptide derived from human SPRY4 (Uniprot #: Q9C004) | |
| 50 μg | |
| Growth and Development, Neuronal Cell Markers, Signal Transduction | |
| 81848 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction