Learn More
Spreader Blue Polystyrene Wedge-Shaped Spreader
High impact S-shape disposable bacterial spreaders
Marke: Microspec D6
Code : 03
Weitere Details : Weight : 0.74700kg
Beschreibung
High impact polystyrene disposable bacterial spreaders. Wedge shaped to reduce tendency to flip, or traditional “L” shape. Packs of 500 contain re-sealable inner bags of 5 sterile spreaders.Spezifikation
Yes | |
L-Shape | |
Blue | |
Polystyrene |
Sterile | |
Spreader | |
100 bags of 5 | |
High impact Polystyrene disposable bacterial spreaders |