Learn More
uvex Sportstyle Glasses
Product Code. 31910418 Shop All Uvex Products
Change view
Click to view available options
Lens Tint:
Silver, Mirror
Unit Size:
Each
Product Code. Lens Tint unitSize
31910418 Silver, Mirror Each
1 options
Product Code. 31910418 Supplier uvex Supplier No. 9193888
  • ONLINE EXCLUSIVE PRICE
    £23.77
    £21.83 / Pack of 1
    Save £1.94
    8% Off
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
Modern eye protection. uvex sportstyle represents a new generation of innovative safety spectacles: A clean, sporty design combining maximum functionality and protection - everyone will find these lightweight safety glasses a pleasure to wear.
  • Sporty, lightweight safety spectacles with contoured lens for a great fit on narrow or wider faces
  • Robust polycarbonate lens with proven uvex supra-vision coating technology - permanently anti-fog and scratch-resistant lenses
  • Combination of soft padding around the brow and ergonomically shaped side arms with soft ear-pieces ensures a comfortable, secure fit without creating pressure points
  • Soft, adjustable nose piece allows spectacles to be adapted to the wearer to deliver a comfortable, non-slip and pressure-free fit
  • Certified in accordance with EN 166 (personal eye protection) and EN 172 (sunglare filters for industrial use) UV400 - 100% protection against harmful UV radiation to 400 nm
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant, Anti-scratch
Lens Tint Silver, Mirror
Product Type Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material TPE
UV Protection UV400
Frame Type Semi-rimless