missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Splicing factor, arginine/serine-rich 11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57324
This item is not returnable.
View return policy
Description
Splicing factor, arginine/serine-rich 11 Polyclonal specifically detects Splicing factor, arginine/serine-rich 11 in Human samples. It is validated for Western Blot.
Specifications
| Splicing factor, arginine/serine-rich 11 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Arginine-rich 54 kDa nuclear protein, dJ677H15.2, DKFZp686M13204, p54NET2, serine/arginine-rich splicing factor 11, SFRS11, splicing factor p54, Splicing factor, arginine/serine-rich 11FLJ18226, SR splicing factor 11 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9295 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q05519 | |
| SRSF11 | |
| Synthetic peptides corresponding to SFRS11(splicing factor, arginine/serine-rich 11) The peptide sequence was selected from the C terminal of SFRS11. Peptide sequence KKKSKDKEKDRERKSESDKDVKQVTRDYDEEEQGYDSEKEKKEEKKPIET. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Chicken: 100%; Human: 100%; Mouse: 92%; Rat: 92%; Bovine: 92%; Pig: 92%;. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction