missing translation for 'onlineSavingsMsg'
Learn More

Spironolactone, Thermo Scientific™

£26.90 - £192.00

Chemical Identifiers

CAS 52-01-7
Molecular Formula C24H32O4S
Molecular Weight (g/mol) 416.576
MDL Number MFCD00082250
InChI Key LXMSZDCAJNLERA-ZHYRCANASA-N
PubChem CID 5833
ChEBI CHEBI:9241
IUPAC Name S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate
SMILES CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15402949
SDS View Documents
Thermo Scientific Alfa Aesar
J60119.03
1g
£26.90
1g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15412949
SDS View Documents
Thermo Scientific Alfa Aesar
J60119.06
5g
£108.00
5g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15422949
SDS View Documents
Thermo Scientific Alfa Aesar
J60119.09
10g
£192.00
10g
View alternative products
Save Up to 
»
Estimated Shipment: 13-10-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in chloroform and methanol. Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

52-01-7
416.576
LXMSZDCAJNLERA-ZHYRCANASA-N
CHEBI:9241
CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C
C24H32O4S
MFCD00082250
5833
S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate

Specifications

52-01-7
C24H32O4S
14,8760
LXMSZDCAJNLERA-ZHYRCANASA-N
S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate
5833
416.58
White
MFCD00082250
Soluble in chloroform and methanol. Slightly soluble in water.
CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C
416.576
CHEBI:9241
Spironolactone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H302
May damage fertility or the unborn child.
Harmful if swallowed.

P201-P202-P260-P281-P308+P313-P501c

H351-H360F-H373

missing translation for 'einecsNumber' : 200-133-6

missing translation for 'rtecsNumber' : TU4725000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Spironolactone

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A

Hazard Statement
  • H302-Harmful if swallowed.
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.