missing translation for 'onlineSavingsMsg'
Learn More

Spironolactone

Product Code. 15422949
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15422949 10 g 10g
15402949 1 g 1g
15412949 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15422949 Supplier Thermo Scientific Alfa Aesar Supplier No. J60119.09
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52-01-7 | C24H32O4S | 416.576 g/mol

Spironolactone used as potassium sparing diuretic. Also used primarily to treat ascites in patients with liver disease, low-renin hypertension, hypokalemia, and Conn’s syndrome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform and methanol. Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-01-7
Molecular Formula C24H32O4S
Molecular Weight (g/mol) 416.576
MDL Number MFCD00082250
InChI Key LXMSZDCAJNLERA-ZHYRCANASA-N
PubChem CID 5833
ChEBI CHEBI:9241
IUPAC Name S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate
SMILES CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C

Specifications

Type Spironolactone
Physical Form Powder
Melting Point 207°C to 208°C
Color White
Quantity 10 g
Merck Index 14,8760
Solubility Information Soluble in chloroform and methanol. Slightly soluble in water.
Formula Weight 416.58
Chemical Name or Material Spironolactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.