missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPINK5 Recombinant Rabbit Monoclonal Antibody (23GB4180), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553705
This item is not returnable.
View return policy
Description
Lympho-epithelial Kazal-type inhibitor (LEKTI) is a serine protease inhibitor which protects mucous epithelia against microbial attack and inflammation. LEKTI is a marker of epithelial differentiation and expresses strongly in the granular and uppermost spinous layers of the epidermis and differentiated layers of stratified epithelia. Defects in SPINK5, the gene encoding LEKTI, are the cause of Netherton syndrome, a severe autosomal recessive disorder characterized by atopic dermatitis, hayfever and other conditions.
Specifications
| SPINK5 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SPINK5 | |
| 2310065D10Rik; AV238912; Hemofiltrate peptide HF6478; Hemofiltrate peptide HF7665; ISK5; LEKT1; LEKTI; LETK1; LETKI; lympho-epithelial Kazal-type-related inhibitor; lymphoepithelial Kazal-type-related inhibitor; Netherton syndrome; NETS; NS; serine peptidase inhibitor, Kazal type 5; serine protease inhibitor Kazal-type 5; serine protease inhibitor, Kazal type 5; SPINK5; VAKT1; VAKTI | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11005, 361319, 72432 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4180 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NQ38 | |
| SPINK5 | |
| A synthesized peptide derived from human SPINK5/LEKTI (150-200AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction