missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPINK5 Recombinant Rabbit Monoclonal Antibody (23GB4180), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | SPINK5 |
|---|---|
| Clone | 23GB4180 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089680
![]() |
Invitrogen™
MA553704 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088682
|
Invitrogen™
MA553705 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPINK5 Recombinant Monoclonal antibody specifically detects SPINK5 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| SPINK5 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q9NQ38 | |
| 11005, 361319, 72432 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4180 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2310065D10Rik; AV238912; Hemofiltrate peptide HF6478; Hemofiltrate peptide HF7665; ISK5; LEKT1; LEKTI; LETK1; LETKI; lympho-epithelial Kazal-type-related inhibitor; lymphoepithelial Kazal-type-related inhibitor; Netherton syndrome; NETS; NS; serine peptidase inhibitor, Kazal type 5; serine protease inhibitor Kazal-type 5; serine protease inhibitor, Kazal type 5; SPINK5; VAKT1; VAKTI | |
| A synthesized peptide derived from human SPINK5/LEKTI (150-200AA). | |
| Primary | |
| -20°C | |
| SPINK5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title