missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPINK5 Monoclonal Antibody (C10), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA543567

 View more versions of this product

Product Code. 17634026

  • £178.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Lympho-epithelial Kazal-type inhibitor (LEKTI) is a serine protease inhibitor which protects mucous epithelia against microbial attack and inflammation. LEKTI is a marker of epithelial differentiation and expresses strongly in the granular and uppermost spinous layers of the epidermis and differentiated layers of stratified epithelia. Defects in SPINK5, the gene encoding LEKTI, are the cause of Netherton syndrome, a severe autosomal recessive disorder characterized by atopic dermatitis, hayfever and other conditions.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPINK5
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q9NQ38
SPINK5
Gly699-Ser976 (Accession Q9NQ38), with N-terminal His Tag.
20 μL
Primary
Human
Antibody
IgG1 κ
Western Blot
C10
Unconjugated
SPINK5
2310065D10Rik; AV238912; Hemofiltrate peptide HF6478; Hemofiltrate peptide HF7665; ISK5; LEKT1; LEKTI; LETK1; LETKI; lympho-epithelial Kazal-type-related inhibitor; lymphoepithelial Kazal-type-related inhibitor; Netherton syndrome; NETS; NS; serine peptidase inhibitor, Kazal type 5; serine protease inhibitor Kazal-type 5; serine protease inhibitor, Kazal type 5; SPINK5; VAKT1; VAKTI
Mouse
Protein A/G
RUO
11005
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.