missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPINK4 Rabbit anti-Human, Clone: 285, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | SPINK4 |
|---|---|
| Clone | 285 |
| Dilution | Sandwich ELISA Capture 1:250-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18231408
|
Novus Biologicals
NBP2-89955-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18204087
|
Novus Biologicals
NBP2-89955-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPINK4 Monoclonal specifically detects SPINK4 in Human samples. It is validated for Sandwich ELISA Capture.Specifications
| SPINK4 | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 27290 | |
| IgG | |
| Protein A purified |
| 285 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| gastrointestinal peptide, MGC133107, serine peptidase inhibitor, Kazal type 4, serine protease inhibitor Kazal-type 4, serine protease inhibitor, Kazal type 4 | |
| Recombinant Human SPINK4 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit