missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPINK4 Rabbit anti-Human, Clone: 285, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89955-100UL
This item is not returnable.
View return policy
Description
SPINK4 Monoclonal specifically detects SPINK4 in Human samples. It is validated for Sandwich ELISA Capture.
Specifications
| SPINK4 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 27290 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 285 | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| gastrointestinal peptide, MGC133107, serine peptidase inhibitor, Kazal type 4, serine protease inhibitor Kazal-type 4, serine protease inhibitor, Kazal type 4 | |
| Recombinant Human SPINK4 Protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction