missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sphingosine Kinase 2/SPHK2 Antibody (3C8D3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £453.00
Specifications
| Antigen | Sphingosine Kinase 2/SPHK2 |
|---|---|
| Clone | 3C8D3 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18244610
|
Novus Biologicals
NBP2-52566 |
0.1 mg |
£453.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692938
|
Novus Biologicals
NBP2-52566-0.025mg |
0.025 mg |
£173.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sphingosine Kinase 2/SPHK2 Monoclonal specifically detects Sphingosine Kinase 2/SPHK2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Sphingosine Kinase 2/SPHK2 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis | |
| PBS with 0.05% Sodium Azide | |
| 56848 | |
| Synthesized peptide of human SK2 (AA: 36-52). | |
| Primary | |
| 69.2 kDa |
| 3C8D3 | |
| Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.1.91, SK 2, sphingosine kinase 2, sphingosine kinase type 2 isoform, SPK 2 | |
| SPHK2 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title