missing translation for 'onlineSavingsMsg'
Learn More

Sphingosine Kinase 2/SPHK2 Antibody (3C8D3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-52566-0.025mg

 View more versions of this product

Product Code. 18692938

  • £173.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sphingosine Kinase 2/SPHK2 Monoclonal specifically detects Sphingosine Kinase 2/SPHK2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sphingosine Kinase 2/SPHK2
Monoclonal
1 mg/mL
Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready
EC 2.7.1.91, SK 2, sphingosine kinase 2, sphingosine kinase type 2 isoform, SPK 2
Mouse
69.2 kDa
0.025 mg
Apoptosis
56848
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
3C8D3
Unconjugated
PBS with 0.05% Sodium Azide
SPHK2
Synthesized peptide of human SK2 (AA: 36-52).
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.