missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Sphingosine Kinase 1/SPHK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £532.00
Specifications
| Antigen | Sphingosine Kinase 1/SPHK1 |
|---|---|
| Dilution | Western Blot : 1:400, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067113
|
Novus Biologicals™
NBP3-30856-100ug |
100 μg |
£532.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066968
|
Novus Biologicals™
NBP3-30856-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sphingosine Kinase 1/SPHK1 Polyclonal antibody specifically detects Sphingosine Kinase 1/SPHK1 in Bovine,Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Sphingosine Kinase 1/SPHK1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| Phosphate buffered saline | |
| 8877 | |
| IgG | |
| Antigen Affinity-purified |
| Western Blot : 1:400, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine, Human, Mouse, Rat | |
| EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1 | |
| Rabbits were immunized with a synthetic peptide derived from the C-terminal of human Sphingosine Kinase 1/SPHK1 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title