missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Sphingosine Kinase 1/SPHK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30856-25ug
This item is not returnable.
View return policy
Description
Sphingosine Kinase 1/SPHK1 Polyclonal antibody specifically detects Sphingosine Kinase 1/SPHK1 in Bovine,Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Sphingosine Kinase 1/SPHK1 | |
| Polyclonal | |
| Western Blot : 1:400, ELISA | |
| EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1 | |
| Rabbits were immunized with a synthetic peptide derived from the C-terminal of human Sphingosine Kinase 1/SPHK1 | |
| 25 μg | |
| Lipid and Metabolism | |
| 8877 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction