missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Sphingomyelin Synthase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £532.00
Specifications
| Antigen | Sphingomyelin Synthase 2 |
|---|---|
| Dilution | Western Blot : 5-10 μg/mL, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067027
|
Novus Biologicals™
NBP3-30870-100ug |
100 μg |
£532.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067167
|
Novus Biologicals™
NBP3-30870-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sphingomyelin Synthase 2 Polyclonal antibody specifically detects Sphingomyelin Synthase 2 in Human samples. It is validated for ELISA,Western BlotSpecifications
| Sphingomyelin Synthase 2 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| EC 2.7.8.27, phosphatidylcholine:ceramide cholinephosphotransferase 2, SM synthase, sphingomyelin synthase 2SMS2MGC26963 | |
| Synthetic peptide derived from the human Sphingomyelin Synthase 2 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 5-10 μg/mL, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline, pH 7.4 | |
| 166929 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title