missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Sphingomyelin Synthase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30870-100ug
This item is not returnable.
View return policy
Description
Sphingomyelin Synthase 2 Polyclonal antibody specifically detects Sphingomyelin Synthase 2 in Human samples. It is validated for ELISA,Western Blot
Specifications
| Sphingomyelin Synthase 2 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL, ELISA | |
| EC 2.7.8.27, phosphatidylcholine:ceramide cholinephosphotransferase 2, SM synthase, sphingomyelin synthase 2SMS2MGC26963 | |
| Synthetic peptide derived from the human Sphingomyelin Synthase 2 protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline, pH 7.4 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 166929 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction