missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPG7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | SPG7 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613540
|
Novus Biologicals
NBP2-93364-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18667300
|
Novus Biologicals
NBP2-93364-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPG7 Polyclonal antibody specifically detects SPG7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SPG7 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neurodegeneration, Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 6687 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| CARMGC126331, cell matrix adhesion regulator, CMARMGC126332, EC 3.4.24, EC 3.4.24.-, FLJ37308, paraplegin, PGNcell adhesion regulator, spastic paraplegia 7 (pure and complicated autosomal recessive), Spastic paraplegia 7 protein, SPG5C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human SPG7 (NP_003110.1). MAVLLLLLRALRRGPGPGPRPLWGPGPAWSPGFPARPGRGRPYMASRPPGDLAEAGGRALQSLQLRLLTPTFEGINGLLLKQHLVQNPVRLWQLLGGTFYFNTSRLKQKNKEKDKSKGKAPEEDEEERRRRERDDQMYRERLRTLLVIAVVMSLLNALSTSGGSISWNDFVHEMLAKGEVQRVQVVPESDVVEVYLHPGAVVFGRPRLALMYRMQVANIDKFEEKLRAAEDELNIEAKDRIPVSYKRTGF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title