missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPG7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93364-0.1ml
This item is not returnable.
View return policy
Description
SPG7 Polyclonal antibody specifically detects SPG7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SPG7 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| CARMGC126331, cell matrix adhesion regulator, CMARMGC126332, EC 3.4.24, EC 3.4.24.-, FLJ37308, paraplegin, PGNcell adhesion regulator, spastic paraplegia 7 (pure and complicated autosomal recessive), Spastic paraplegia 7 protein, SPG5C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human SPG7 (NP_003110.1). MAVLLLLLRALRRGPGPGPRPLWGPGPAWSPGFPARPGRGRPYMASRPPGDLAEAGGRALQSLQLRLLTPTFEGINGLLLKQHLVQNPVRLWQLLGGTFYFNTSRLKQKNKEKDKSKGKAPEEDEEERRRRERDDQMYRERLRTLLVIAVVMSLLNALSTSGGSISWNDFVHEMLAKGEVQRVQVVPESDVVEVYLHPGAVVFGRPRLALMYRMQVANIDKFEEKLRAAEDELNIEAKDRIPVSYKRTGF | |
| 0.1 mL | |
| Cell Biology, Neurodegeneration, Neuroscience | |
| 6687 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction