Learn More
Astell Scientific™ SPF Seal
Description
- Acts as a barrier to prevent bacterial migration.
- Used in double-door systems (loading, non-sterile/unloading, sterile).
Specifications
Specifications
| For Use With (Equipment) | Double-Door Autoclave Systems |