Learn More
Cole-Parmer™ Grinding Ball Dispenser for 96-Well Microplates
Product Code. 12869045 Shop All SPEX Certiprep Products
Change view
Click to view available options
For Use With (Equipment):
Grinding Ball and 96-Well Titer Plates of Geno/Grinder
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
12869045 Grinding Ball and 96-Well Titer Plates of Geno/Grinder Each
1 options
Product Code. 12869045 Supplier Cole-Parmer™ Supplier No. 2100SSP
Grinding Ball Dispenser for use with Geno/Grinder™.
  • Loads each well of a standard 96-well titer plate with one grinding ball in less than a second.
Specifications
Product Type Grinding Ball Dispenser
For Use With (Equipment) Grinding Ball and 96-Well Titer Plates of Geno/Grinder
Holds 96-well Microplates
Material Stainless Steel (440 C)
Packaging Quantity 1 Ea.