Learn More
Cole-Parmer™ Replacement End Plugs for Large Grinding Vial Set, Stainless Steel
Shop All SPEX Certiprep Products
Click to view available options
Packaging Quantity:
2 Pk.
Unit Size:
Pack of 2
Description
- Made of 440C stainless steel.
- Withstands liquid nitrogen temperatures.
- Autoclavable and reusable.
Specifications
Specifications
| Product Type | End Plug |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| For Use With (Equipment) | 6801 Large Grinding Vial Set |
| Material | Stainless Steel (440 C) |
| Packaging Quantity | 2 Pk. |