Learn More
SPEX™ SamplePrep Large-Capacity Freezer/Mill™ Auto-Fill System
Shop All SPEX Certiprep Products
Click to view available options
:
Auto-fill system
Description
- Automatically maintains proper level of liquid nitrogen in 6850 mill during operation
- Provides safe, efficient liquid nitrogen handling
- Minimizes fluid loss, saves refill time
- Requires cryogenic transfer hose (not included; contact your Fisher Scientific Sales Representative for details)
Specifications
Specifications
| Product Type | Autofill System |
| For Use With (Equipment) | Spex SamplePrep Large Capacity Freezer/Mill |
| Includes | Adapter, short cryogenic transfer hose, long cryogenic transfer hose |