Learn More
SPEX™ SamplePrep Cryo-Adapter for 2210 Titer Plates
Shop All SPEX Certiprep Products
Click to view available options
For Use With (Equipment):
2210 Titer Plate
Unit Size:
Each
Description
- Cryo-Blocks are chilled in the 2600 Cryo-Station and then used for high-throughput cryogenic grinding
- Enables high-throughput cryogenic grinding in the Geno/Grinder™
- Maintains samples at liquid nitrogen temperatures during tissue homogenization and cell lysis
- Preserves pesticides, RNA, and proteins for heat sensitive extraction methods
- Increases yields by grinding samples at sub-zero temperatures
- Used two at a time in the Standard Clamp Assembly to keep it in balance
Specifications
Specifications
| Product Type | Cryo Adapter |
| For Use With (Application) | Ideal for High Throughput Cryogenic Grinding |
| For Use With (Equipment) | 2210 Titer Plate |
| Material | Aluminum |