Learn More
Molecular Devices SpectraMax™ M5 Cuvette/Microplate Reader with SoftMax Pro Software
Shop All Molecular Devices ProductsDescription
- Designed for multi-mode detection workflows in microplate- and cuvette-based assays
- Supports absorbance, fluorescence, and luminescence measurement applications
- Enables flexible assay setup with monochromator-based wavelength selection
- Supports biochemical and cell-based assay workflows across life science applications
- Supports both microplate and cuvette formats for expanded experimental flexibility
- Supports endpoint, kinetic, and spectral measurement workflows
- Provides configurable detection settings for diverse assay requirements
- Integrated with SoftMax Pro Software for data acquisition and analysis
Specifications
Specifications
| Incubator Temperature | 4-Zone Incubation to 45°C |
| Measurement Speed | 11 sec. (96 wells), 22 sec. (384 wells) |
| Shaking | Linear, Orbital |
| Plate Type | 6 to 384-well Plates |
| Detector Type | PMT |
| Software Type | SoftMax Pro |
| Light Source | Xenon Flash |
| Reading Options | Endpoint, Kinetic, Spectral Scanning |