Learn More
Molecular Devices SpectraMax™ L Microplate Reader with SoftMax Pro Software
Product Code. 31936374 Shop All Molecular Devices Products
Change view
Click to view available options
Plate Type:
96 and 384-well Plates
Unit Size:
1 set
Product Code. Plate Type Unit Size
31936374 96 and 384-well Plates 1 set
1 options
Product Code. 31936374 Supplier Molecular Devices Supplier No. SPECTRAMAX L CONFIG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Perform sensitive luminescence detection for microplate‐based assays. Supports luminescence workflows for a wide range of life science applications.
  • Designed for luminescence detection workflows in microplate-based assays
  • Supports luminescence measurement applications for life science research
  • Enables flexible assay setup with configurable detection parameters
  • Supports biochemical and cell-based assay workflows across life science applications
  • Provides programmable measurement workflows for routine and advanced assays
  • Supports microplate-based screening applications across multiple formats
  • Integrated with SoftMax Pro Software for data acquisition and analysis
  • Designed for sensitive detection of luminescent assay signals
TRUSTED_SUSTAINABILITY
Specifica
Detector Type Photomultiplier Tube (PMT)
Interface PC Control, Internal Software
Light Source LED
Measurement Technology Luminescence (Top)
Plate Type 96 and 384-well Plates
Reading Options Endpoint
Shaking None
Wavelength Selection Filters (FL, Lum.)
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.