Learn More
Molecular Devices SpectraMax™ iD5e MultiMode Platform IN‐AS with SoftMax Pro Software
Shop All Molecular Devices ProductsDescription
- Designed for multi-mode detection workflows in microplate-based assays
- Supports a broad range of detection applications for complex assay workflows
- Enables flexible assay setup with tunable monochromator- and filter-based detection
- Supports biochemical and cell-based assay workflows across life science applications
- Includes integrated fluid handling capability for automated reagent addition
- Provides enhanced detection capability for advanced and evolving assay requirements
- Includes configuration combining IN and AS platform options for flexible workflow setup
- Includes preconfigured protocols to support common assay workflows
- Integrated with SoftMax Pro Software for data acquisition and analysis
Specifications
Specifications
| Incubator Temperature | 4-Zone Incubation to 70°C |
| Measurement Speed | 11 sec. (96 wells), 22 sec. (384 wells) |
| Shaking | Linear, Orbital, Double Orbital |
| Plate Type | 6 to 384-well Plates |
| Detector Type | PMT |
| Software Type | SoftMax Pro |
| Light Source | Xenon Flash |
| Reading Options | Endpoint, Kinetic, Spectral Scanning |